missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-3-furaldehyde, 97%, Thermo Scientific™

670.00 SEK - 1740.00 SEK

Chemical Identifiers

CAS 157836-53-8
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD08690281
InChI Key KWEAOVDZILYEET-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furaldehyde, 3-furancarboxaldehyde,5-methyl-2-phenyl
PubChem CID 10397534
IUPAC Name 5-methyl-2-phenylfuran-3-carbaldehyde
SMILES CC1=CC(C=O)=C(O1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10032404
View Documents
Thermo Scientific Maybridge
250 mg Amber Glass Bottle
670.00 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
10798065
View Documents
Thermo Scientific Maybridge
1 g Amber Glass Bottle
1740.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

157836-53-8
186.21
KWEAOVDZILYEET-UHFFFAOYSA-N
10397534
CC1=CC(C=O)=C(O1)C1=CC=CC=C1
C12H10O2
MFCD08690281
5-methyl-2-phenyl-3-furaldehyde, 3-furancarboxaldehyde,5-methyl-2-phenyl
5-methyl-2-phenylfuran-3-carbaldehyde

Specifications

157836-53-8
100.0
C12H10O2
5-methyl-2-phenyl-3-furaldehyde, 3-furancarboxaldehyde,5-methyl-2-phenyl
CC1=CC(C=O)=C(O1)C1=CC=CC=C1
186.21
186.21
5-Methyl-2-phenyl-3-furaldehyde
96.5
97%
MFCD08690281
KWEAOVDZILYEET-UHFFFAOYSA-N
5-methyl-2-phenylfuran-3-carbaldehyde
10397534
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications