missing translation for 'onlineSavingsMsg'
Learn More

5-Methylimidazole-4-carboxaldehyde, 99%, Thermo Scientific Chemicals

419.00 SEK - 1435.00 SEK

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zsShow MoreShow Less
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479027
SDS View Documents
Thermo Scientific Alfa Aesar
B20608.03
1 g N/A N/A N/A
11489027
SDS View Documents
Thermo Scientific Alfa Aesar
B20608.06
5 g
1435.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.

Solubility
Soluble in water, (23 g/L) (25°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methylimidazole-4-carboxaldehyde, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.