Learn More
5-tert-Butyl-m-xylene, 98%, Thermo Scientific Chemicals
388.00 SEK - 1195.00 SEK
Chemical Identifiers
CAS | 98-19-1 |
---|---|
Molecular Formula | C12H18 |
Molecular Weight (g/mol) | 162.276 |
MDL Number | MFCD00008832 |
InChI Key | FZSPYHREEHYLCB-UHFFFAOYSA-N |
Synonym | 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl |
PubChem CID | 7378 |
IUPAC Name | 1-tert-butyl-3,5-dimethylbenzene |
SMILES | CC1=CC(=CC(=C1)C(C)(C)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449496
|
Thermo Scientific Alfa Aesar
A19220.22 |
100 g |
388.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459496
|
Thermo Scientific Alfa Aesar
A19220.36 |
500 g |
1195.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-19-1 | |
162.276 | |
FZSPYHREEHYLCB-UHFFFAOYSA-N | |
7378 | |
CC1=CC(=CC(=C1)C(C)(C)C)C |
C12H18 | |
MFCD00008832 | |
5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
1-tert-butyl-3,5-dimethylbenzene |
Specifications
98-19-1 | |
0.86 | |
84°C (183°F) | |
1.496 | |
1853314 | |
FZSPYHREEHYLCB-UHFFFAOYSA-N | |
1-tert-butyl-3,5-dimethylbenzene | |
7378 | |
98% |
-22°C | |
207°C to 209°C | |
C12H18 | |
MFCD00008832 | |
5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
CC1=CC(=CC(=C1)C(C)(C)C)C | |
162.276 | |
162.27 | |
5-tert-Butyl-m-xylene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 202-647-6
missing translation for 'rtecsNumber' : ZE3900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures