- Packaged under Argon
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
G16W001 | Certificate of Analysis | 24/11/2021 | 045913.MD,045918.06,045913.03 |
Z01A017 | Certificate of Analysis | 22/11/2021 | 045918.06,045921.06,045918.14 |
I07X034 | Certificate of Analysis | 22/11/2021 | 045918.06,045921.06,045918.14 |
Q09H034 | Certificate of Analysis | 22/11/2021 | 045918.06,045921.06,045918.14 |
RUO – Research Use Only
Product Content Correction
Product Title
Dysprosium powder, -200 mesh, 99.9% (REO), Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction