Learn More
Tocris Bioscience™ 5J 4
CRAC channel blocker; anti-inflammatory
Brand: Tocris Bioscience™ 5490/50
5636.14 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.Specifications
N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide | |
50 mg | |
C16H12N2O3S | |
>98% |
827001-82-1 | |
Other Calcium Channel Blockers | |
312.34 |