missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole, 98%, Thermo Scientific Chemicals

Product Code. 11378646
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52415-29-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00238550
InChI Key MAWGHOPSCKCTPA-UHFFFAOYSA-N
Synonym 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
PubChem CID 676493
IUPAC Name 6-bromo-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)Br

Specifications

Melting Point 91°C to 93°C
Boiling Point 70°C to 75°C (0.01 mmHg)
Quantity 250 mg
Beilstein 112711
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 6-Bromoindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'