missing translation for 'onlineSavingsMsg'
Learn More
6-Chloro-1-hexene, 97%, Thermo Scientific Chemicals Product Code.: 15430168

6-Chloro-1-hexene, 97%, Thermo Scientific Chemicals

Product Code. 15430168
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15430168

Brand: Thermo Scientific Alfa Aesar H53396.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-Chloro-1-hexene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-89-2
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00039391
InChI Key BLMIXWDJHNJWDT-UHFFFAOYSA-N
Synonym 6-chloro-1-hexene, 1-hexene, 6-chloro, 5-hexenyl chloride, 6-chlorohexene, chloro-1-hex, 6-chloro-1-hexen, 1-chloro-5-hexene, 1-chlorohex-5-ene, acmc-20alz4, 5-hexen-1-yl chlorideShow MoreShow Less
PubChem CID 70233
IUPAC Name 6-chlorohex-1-ene
SMILES C=CCCCCCl

Specifications

Density 0.896
Boiling Point 135°C to 136°C
Flash Point 29°C (84°F)
Linear Formula Cl(CH2)4CH=CH2
Refractive Index 1.438
Quantity 25 g
UN Number UN1993
Formula Weight 118.61
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-hexene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW23K018 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberH53396.06,H53396.14
Lot NumberK19W109 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53396.06,H53396.14
Lot NumberF19Z003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53396.06,H53396.14
Lot NumberT19F019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53396.06,H53396.14
Lot NumberX05B028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53396.06,H53396.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-1-hexene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.