missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-2-fluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air and reducing agents. Store under an inert atmosphere.
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air and reducing agents. Store under an inert atmosphere.

Specifications
Specifications
Melting Point | 95°C to 100°C |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Formula Weight | 188.59 |
Percent Purity | 97% |
Chemical Name or Material | 6-Chloro-2-fluoro-3-methoxybenzaldehyde |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-Chloro-2-fluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction