missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals

709.00 SEK - 11990.00 SEK

Chemical Identifiers

CAS 1191-25-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00046560
InChI Key IWHLYPDWHHPVAA-UHFFFAOYSA-N
Synonym 6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
PubChem CID 14490
ChEBI CHEBI:17869
IUPAC Name 6-hydroxyhexanoic acid
SMILES C(CCC(=O)O)CCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11476646
View Documents
Thermo Scientific Alfa Aesar
B24857.03
1 g
709.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11486646
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24857.06
5 g
2800.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11496646
View Documents
Thermo Scientific Alfa Aesar
B24857.14
25 g
11990.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1191-25-9
132.159
IWHLYPDWHHPVAA-UHFFFAOYSA-N
14490
6-hydroxyhexanoic acid
C6H12O3
MFCD00046560
6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
CHEBI:17869
C(CCC(=O)O)CCO

Specifications

1191-25-9
1.08
C6H12O3
1.456
6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acid
C(CCC(=O)O)CCO
132.159
CHEBI:17869
95%
6-Hydroxyhexanoic acid
38°C to 40°C
>110°C (230°F)
HO(CH2)5CO2H
MFCD00046560
IWHLYPDWHHPVAA-UHFFFAOYSA-N
6-hydroxyhexanoic acid
14490
132.16
may cont. variable amounts of dimer
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only