missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-O-α-Maltosyl-β-cyclodextrin, MedChemExpress
MedChemExpress 6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.
Brand: Medchem Express HY-18593

This item is not returnable.
View return policy
Description
6-O-α-Maltosyl-β-cyclodextrin is a cellular cholesterol modifier which can form soluble inclusion complex with cholesterol.Specifications
6-O-α-Maltosyl-β-cyclodextrin | |
White | |
C54H90O45 | |
6-O-alpha-D-Maltosyl-beta-cyclodextrin | |
O[C@@H]1[C@@H](CO)O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](OC[C@H]3O[C@H](O[C@@H]4[C@@H](CO)O[C@H](O[C@@H]5[C@@H](CO)O[C@H](O6)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@@H](O)[C@@H]3O[C@H]7O[C@H](CO)[C@@H](O[C@@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]9[C@H](O)[C@@H](O)[C@H](OC%10[C@H](O)[C@@H](O)[C@H]6[C@@H](CO)O%10)[C@@H](CO)O9)[C@@H](CO)O8)[C@H](O)[C@H]7O)O[C@@H]2CO)[C@H](O)[C@H]1O | |
1459.27 | |
Research |
104723-60-6 | |
Research | |
5 g | |
DMSO : ≥ 100 mg/mL (68.53 mM) ∣H2O : ≥ 60.5 mg/mL (41.46 mM) | |
1459.27 | |
97.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
6-O-α-Maltosyl-β-cyclodextrin, MedChemExpress
Research purposes only
Spot an opportunity for improvement?Share a Content Correction