missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Product Code. 15406246
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This compounds are useful laboratory reagents and used frequently in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compounds are useful laboratory reagents and used frequently in organic synthesis.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122138-83-4
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.153
MDL Number MFCD23379858
InChI Key PEOVRVSERLAIQM-UHFFFAOYSA-N
PubChem CID 14422956
IUPAC Name 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F

Specifications

Quantity 1 g
Formula Weight 198.15
Percent Purity 97%
Chemical Name or Material 7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'