Learn More
Thermo Scientific™ 7500FAST Adaptor
Applied Biosystems 7500 Fast 96W Strip Plate Adaptor.
Brand: Thermo Scientific™ A46347
1924.37 SEK valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.05000kg
Description
- 7500 Fast 96W Strip Plate Adaptor
- Accessory
Specifications
SureTect | |
SureTect PCR Tool |