Learn More
Brady™ 7942 Series Thermal Transfer Printer Ribbon
General purpose wax ribbon for paper or temporary labeling.
Brand: Brady™ 804447
218.74 SEK valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
General purpose, standard performance wax ribbon for paper and temporary labeling.
Specifications
Black | |
Wax | |
4.3 in. | |
110 mm | |
12.7 mm | |
2011/65/EU and 2015/863 |
BBP11 Label Printer, BBP12 Label Printer | |
230 ft. | |
70 m | |
0.5 in. | |
Open-core Ribbon |