Learn More
Cytiva 903 Protein Saver Cards
Save time and costs during sample collection, transport, and archiving. Whatman™ 903 Protein Saver Card (EU) fits in the Whatman™ foil barrier resealable bags for storage.
Brand: Cytiva 10531018
2196.40 SEK valid until 2024-12-31
Use promo code "21887" to get your promotional price.
Additional Details : Weight : 0.36000kg
Description
Save time and costs during sample collection, transport, and archiving. Whatman™ 903 Protein Saver Card (EU) fits in the Whatman™ foil barrier resealable bags for storage.
- Ready-to-use cards and kits
- Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity
- The sample collection area of our 903 Proteinsaver Card contains five half-inch circles
- Each circle holds 75-80 μl of sample
- Wraparound cover has spaces for name and date of collection and is imprinted with the universal biohazard symbol
Specifications
Base Scale | |
ISO 9001 |