Learn More
Uvex™ 9301 Ultravision Gas-Tight Goggles
168.99 SEK valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Red
Description
- Comfortable, ergonomically designed full-view goggles
- Suitable for glasses wearers, unrestricted side visibility, problem-free lens change
- Clear, polycarbonate lens with supravision excellence coating
- Gas-tight without a ventilation system
- Heat resistant support body material
Supravision Excellence Coating
- Inside: Anti-fog
- Outside: Scratch-resistant and chemical-resistant
- Easy to clean, and less susceptible to soiling
Specifications
Certifications/Compliance | EN 166:2001, EN 170:2002 |
Lens Type | Supravision Excellence |
Description | Gastight |
Color | Red |
Lens Tint | Clear |
Model | 9301 Ultravision |
Ventilation Details | Not Ventilated |
Disposable | Reusable |
Lens Material | Polycarbonate |
UV Protection | UV400 |