Learn More
uvex 9307 Carbonvision Safety Goggles
Product Code. 15336152
Click to view available options
Lens Tint:
Clear
Gray
Frame Material:
Neoprene
Textile
This item is not returnable. View return policy
This item is not returnable. View return policy
Carbonvision goggles feature a frame that is constructed using the latest materials and manufacturing technologies, for maximum mechanical rigidity (B = 120 m/s) despite minimal volume and weight.
  • Frame is constructed with the latest materials and technologies for maximum mechanical stability, while still remaining lightweight
  • A TPU layer is firmly welded onto the frame to ensure the perfect fit and allow for easy cleaning
  • Certified to EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use)
  • Supravision extreme coating is scratch-proof on the outside, and permanently anti-fog on the inside
  • Offers a panoramic view without a tunnel effect due to the close-fitting lens
  • Ideal for long periods of wear without pain or pressure points
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Supravision Extreme
Lens Coating Anti-fog
Certifications/Compliance (Europe) EN 166:2001, EN 170:2002
Lens Tint Clear
Product Type Anti-fog Goggles
Frame Color Black, Gray
Model 9307 Carbonvision
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Textile
UV Protection UV400
Frame Type Standard
Labeling W 166 34 B CE – 2-1.2 W 1 B KN DIN CE