man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Microplates
Chromatography Microplates
11838981
Print
Fisherbrand™ 96 Well Plate, PP, Square well, Non-sterile, chromatography tested
44 mm height,2000 μL Total Volume,U bottom
Chromatography tested.
Brand:
Fisherbrand™ 08 05 0894
Product Code.
11838981
101.00 SEK
/ Each
Quantity
In Stock
Log in
to see stock.
Explore more special offers
Description
Height: 44mm
2000μL
U bottom
Specifications
Specifications
Format
Standard
Material
Polypropylene
Sterility
Non-sterile
Well Shape
Round Bottom (U-well)
Array
8 x 12
No. of Wells
96
Total Volume (Metric) Well
2000 μL
Height (Metric)
44.3 mm