Learn More
uvex 9906 Pheos Replacement Visor Lens SLB1
Replacement visor lens for pheos series safety helmets is compatible with model 9906.002 (mechanical lever arms), or model 9906.003 (self-locking magnetic catches).
Brand: uvex 9906012
430.15 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement lens for uvex pheos model 9906.002 (mechanical lever arms) or model 9906.003 (self-locking magnetic catches)
- Lightweight, robust face shield for uvex pheos industrial safety helmet ES-WR / BS-WR and uvex pheos alpine (visor adapter 9790.067 required)
- Labeling: 2C-1,2 W1 B 8-1-0 9 KN CE 0196
- Features supravision excellence coating
- Arc flash class 1
Supravision Excellence Coating
- Inside: Anti-fog
- Outside: Scratch-resistant and chemical-resistant
- Easy to clean, and less susceptible to soiling
Specifications
SLB1 | |
Replacement Visor | |
Supravision Excellence |