Learn More
A-867744, MedChemExpress
MedChemExpress A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
Brand: Medchem Express HY-12149
This item is not returnable.
View return policy
Description
A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.Specifications
A-867744 | |
Off-White | |
C20H19ClN2O3S | |
DMSO : ≥ 100 mg/mL (248.21 mM) | |
402.89 | |
95.06% | |
Solid |
1000279-69-5 | |
Research | |
100 mg | |
O=S(C1=CC=C(C=C1)N2C(C)=C(C=C2C3=CC=C(C=C3)Cl)C(CC)=O)(N)=O | |
402.89 | |
Research | |
Neuroscience-Neuromodulation |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
A-867744, MedChemExpress