Learn More
Atago™ Abbe DR-A1 Refractometer
Brand:  Atago™ 1310
Product Code. 10041132
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Specifications
Refractometer
±0.0002nD
With AC adapter
130 x 290 x 310 mm
130 mm
5°C to 50°C
100/240 V
1.3 to 1.7
Digital
1 Each
310 mm
290 mm
100/240 V 50/60 Hz
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products