Learn More
Abemaciclib methanesulfonate, MedChemExpress
MedChemExpress Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Brand:  Medchem Express HY-16297
Encompass_Preferred
 View more versions of this product
Product Code. 16431806
  • 5125.86 SEK / 500mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Specifications
Specifications
Abemaciclib methanesulfonate
Light Yellow
C28H36F2N8O3S
LY2835219 methanesulfonate
CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O
602.7
Research
Cancer-Kinase/protease
1231930-82-7
Research
500 mg
H2O : 125 mg/mL (207.40 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (16.59 mM; ultrasonic and warming and heat to 80°C)
602.7
98.0%
Solid