Learn More
Abemaciclib methanesulfonate, MedChemExpress
MedChemExpress Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Brand: Medchem Express HY-16297
This item is not returnable.
View return policy
Description
Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.Specifications
Abemaciclib methanesulfonate | |
Light Yellow | |
C28H36F2N8O3S | |
LY2835219 methanesulfonate | |
CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O | |
602.7 | |
Research | |
Cancer-Kinase/protease |
1231930-82-7 | |
Research | |
500 mg | |
H2O : 125 mg/mL (207.40 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (16.59 mM; ultrasonic and warming and heat to 80°C) | |
602.7 | |
98.0% | |
Solid |
Research purposes only