Learn More
Abnova™ 2p Subtelomere (Cy5) FISH Probe
Labeled FISH probes for identification of subtelomere aberrations using Fluorescent In Situ Hybridization Technique.
Brand:  Abnova™ FE0015.20uL
Additional Details : Weight : 0.02000kg
Product Code. 16082785
  • 12340.00 SEK / 20µL
Estimated Shipment: 20-08-2024
to see stock.
Includes: FISH Hybridization Buffer (80μL)
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell)
2p Subtelomere
20 μL
Cy5
FISH Hybridization Buffer (80 uL)
2p Subtelomere(Cy5) FISH Probe