missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Lipase Assay Kit
Used for measuring lipase activity and functional study
Specifications
Detection Method | Colorimetric |
---|---|
Type | Lipase Assay Kit |
Includes:
- Lipases assay buffer, 25mL
- OxiRed probe (in DMSO), 0.2mL
- Enzyme mix (lyophilized), 1 vial
- Lipase substrate, 0.4mL
- Glycerol standard (100mM), 0.2mL
- Lipase positive control (lyophilized), 1 vial
Specifications
Colorimetric | |
Lipase Assay Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ Lipase Assay Kit