Learn More
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Optimize performance and flexibility
113.00 SEK - 1805.00 SEK
Specifications
For Use With (Equipment) | For use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers |
---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
15386607
|
Fisherbrand™
88860143 |
Support Rod (12mm dia., 400mm, M10) |
436.00 SEK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
15376607
|
Fisherbrand™
88860142 |
Transparent Shield (PC) |
278.72 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||||||
15366607
|
Fisherbrand™
88860141 |
Heating Bath |
538.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
15336617
|
Fisherbrand™
88860147 |
Temperature Probe (PT 100, SN-8-4 connector sensor), Available for RT2 Advanced Hotplate Stirrer only |
1205.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||||||
15356617
|
Fisherbrand™
88860149 |
3 Prong Clamp (60 mm Grip) |
193.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||||||
15346617
|
Fisherbrand™
88860148 |
C-5, Clamp Holder (PP body, 12mm dia.) |
113.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
15366617
|
Fisherbrand™
88860150 |
Temperature Probe (PT100, A Class, 400C) |
1805.00 SEK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||||||
Description
Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers. Optimize performance and flexibility
- Accessory for use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers
Specifications
For use with RT Hotplate, RT Basic and Advanced Hotplate Stirrers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Isotemp RT Hotplate, RT Basic and Advanced Hotplate Stirrers