Learn More
Cytiva Accessories For Multiphor™ II Electrophoresis Systems
Accessories for analytical or preparative separations of proteins and peptides by isoelectric focusing (IEF), native or SDS PAGE, 2-D PAGE and DNA electrophoresis
Brand: Cytiva 80112939
This item is not returnable.
View return policy
Description
- Assists high quality production of lab-cast gels
Specifications
Preserving | |
Multiphor II Electrophoresis System | |
Mylar Sheet 125 x 250 mm (50/pk.) | |
Complete packsize |
Product Content Correction
Product Title
Cytiva Accessories For Multiphor™ II Electrophoresis Systems > Mylar sheet; Size: 125 x 260mm
Spot an opportunity for improvement?Share a Content Correction