Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units:Glass Plate
Brand: Cytiva SE202P-10
1055.20 SEK valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
SE 250 Mini Vertical Gel Electrophoresis Unit | |
Glass | |
10 cm |
Glass Plate | |
Rectangular | |
8 cm |