Learn More
Thermo Scientific™ Acclaim™ Guard Holder and Coupler
Use the Thermo Scientific™ Guard Holder and Coupler for your Thermo Scientific™ Acclaim™ Guard columns. They can be purchased separately or as a kit.
Brand: Thermo Scientific™ 069707
3685.78 SEK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
Use the Thermo Scientific™ Guard Holder and Coupler for your Thermo Scientific™ Acclaim™ Guard columns. They can be purchased separately or as a kit.
Specifications
Acclaim Guard Cartridge Holder-Coupler Kit V-2 | |
Acclaim | |
Guard column holder and coupler | |
Responsibly packaged |
All Acclaim Guard Cartridges | |
Acclaim Guard Kit (Holder and Coupler) V-2 | |
Each |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
35Hours
5Minutes
22Seconds
58New Offer Save Up to 70%
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.