Learn More
Invitrogen™ AccuPrime™ SuperMix I
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.
4865.00 SEK - 17290.00 SEK
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10564233
|
Invitrogen™
12342010 |
200 Reactions |
4865.00 SEK
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297522
|
Invitrogen™
12342028 |
1000 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix I sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.
In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.
AccuPrime SuperMix I is designed for amplification of genomic DNA amplicons (> 200 bp), plasmid DNA, or cDNA templates. Alternatively, AccuPrime SuperMix II is designed to amplify a broader range of amplicon sizes up to 4 kb.
• Built-in hot start
Order Info
Shipping Condition: Dry Ice
Specifications
Low | |
2X |
For Research Use Only. Not for use in diagnostic procedures.