missing translation for 'onlineSavingsMsg'
Learn More

ACE2 Mouse anti-Human, Clone: 2F12A4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66699-1-IG-150UL

 View more versions of this product

Product Code. 16818825

  • 4721.67 SEK / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody is specific to ACE2.

Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Specifications

Specifications

ACE2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BYF1
ACE2
ACE2 Fusion Protein Ag15554
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2F12A4
Unconjugated
ACE2
ACE related carboxypeptidase, ACE2, ACEH, DKFZp434A014, Metalloprotease MPROT15
Mouse
Protein G
RUO
59272
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers