missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE2 Mouse anti-Human, Clone: 2F12A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66699-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ACE2.
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.Specifications
ACE2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BYF1 | |
ACE2 | |
ACE2 Fusion Protein Ag15554 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2F12A4 | |
Unconjugated | |
ACE2 | |
ACE related carboxypeptidase, ACE2, ACEH, DKFZp434A014, Metalloprotease MPROT15 | |
Mouse | |
Protein G | |
RUO | |
59272 | |
-20°C | |
Liquid |