missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Muscle Monoclonal Antibody (MSA06 (HUC1-1)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511874
This item is not returnable.
View return policy
Description
This antibody reacts with alpha-smooth muscle, alpha-skeletal and alpha-cardiac (sarcomeric). This pan muscle actin antibody reacts with tumors arising from smooth muscle (leiomysarcomas) as well as skeletal muscle tumors (rhabdomyosarcomas).
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.Specifications
Actin Muscle | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P62739, P68138, Q3ZC07, P62736, P68032, P68133, P62737, P68033, P68134, P68137, P62738, P68035, P68136 | |
ACTA1, ACTA2, ACTC1 | |
Purified actin | |
500 μL | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
MSA06 (HUC1-1) | |
Unconjugated | |
ACTA2 | |
actin, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, alpha-sma | |
Mouse | |
Protein G | |
RUO | |
100057678, 100062175, 100065454, 100152267, 100154254, 11459, 11464, 11475, 281592, 29275, 29437, 477587, 478250, 488984, 515610, 533219, 58, 59, 70, 733615, 81633 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction