Learn More
Eppendorf™ Adapter, for 12 tubes 50 mL Nalgene™ Oak Ridge, 2 pcs.
Brand:  Eppendorf™ 5920753002
3363.00 SEK valid until 2024-11-30
Use promo code "21681" to get your promotional price.
Product Code. 16040772
  • 3765.00 SEK / Each
Explore more special offers
Description
Description
Adapter, for 12 tubes 50 mL Nalgene™ Oak Ridge, for Rotor S-4xUniversal-Large, 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapters, Buckets, and Accessories
Rotor S-4 xUniversal-Large
12
2