Learn More
Thermo Scientific™ CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
Shop All Thermo Scientific Temperature Control Solutions Products
Click to view available options
Description:
CentriPAK BPC Adapter
CentriPAK BPC Adapter Rack
CentriPAK BPC Clip
CentriPAK BPC Hanging Rack
Material:
Polyethylene
Polypropylene
Stainless Steel
Autoclavable:
Autoclavable
Not Autoclavable
Description
Accessory for Thermo Scientific™ CentriPAK™ BioProcess Container (BPC). CentriPAK BPC is designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC) enables you to:
- Reduce cross-contamination risk while separating cell cultures
- Perform gentle, high-throughput and high-efficiency harvesting through centrifugation
- Eliminate post-use cleaning steps required with reusable containers
- Meet rigorous quality control and assurance practices for the manufacture of industry-standard material components and aseptic product contact film
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Type | CentriPAK Accessory |
Product Line | Polaris |
Material | Stainless Steel |
Capacity | 6 x 2000 mL, 8 x 2000 mL |
Autoclavable | Not Autoclavable |
For Use With (Equipment) | CentriPAK BioProcess Container |
Description | CentriPAK BPC Adapter Rack |
Length (Metric) | 54.9 cm |
Height (Metric) | 16.2 cm |
Width (Metric) | 21.4 cm |
Show More |
Product Content Correction
Product Title
Thermo Scientific™ CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge >
Spot an opportunity for improvement?Share a Content Correction