Learn More
Hettich™ Adapter for Schlenk Tubes
Designed for use with ROTIXA 500 RS Centrifuge. These rotors are ideal for high sample throughput in medical, research and quality control laboratories.
Brand: Hettich™ 4317
This item is not returnable.
View return policy
Description
- Adapter for Schlenk Tubes
- For use with Rotor no. 4316 and 5616
- For ROTIXA Centrifuges
Specifications
Adapter | |
25 mL | |
146.5 mm | |
1 |
Schlenk | |
24 mm | |
Rotor no. 4316 |