Learn More
Hettich™ Adapter for Schlenk Tubes
Designed for use with ROTIXA 500 RS Centrifuge. These rotors are ideal for high sample throughput in medical, research and quality control laboratories.
Brand:  Hettich™ 4317
Product Code. 12093904
  • 705.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Adapter for Schlenk Tubes
  • For use with Rotor no. 4316 and 5616
  • For ROTIXA Centrifuges
Specifications
Specifications
Adapter
25 mL
146.5 mm
1
Schlenk
24 mm
Rotor no. 4316