Learn More
SCHOTT AG Lighting and Imaging™ Adapter Set for VisiLED Series Illuminators
Combi-kit, adapter set for objective working distances 30 to 110 mm.
Brand: SCHOTT AG Lighting and Imaging™ 400500
This item is not returnable.
View return policy
Description
- Adapter Set to mount VisiLed Darkfield Ring Light (400 300) to VisiLed Brightfield Ring Light (400255, 400225 or 400 100).
Specifications
Adapter Kit | |
Brightfield, Darkfield Adapter Kit | |
Stereo Microscopy Illumination | |
VisiLED Darkfield and Brightfield Ringlight |
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ Adapter Set for VisiLED Series Illuminators > Adapter Set for VisiLED Series Illuminators; For Use With: VisiLED Ringlights S80-55, S40-55, S40-10D
Spot an opportunity for improvement?Share a Content Correction