missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCK5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-84398-0.1ML
This item is not returnable.
View return policy
Description
ADCK5 Polyclonal specifically detects ADCK5 in Mouse samples. It is validated for Western Blot.Specifications
ADCK5 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
aarF domain containing kinase 5, EC 2.7.11.-, FLJ35454, MGC126708 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse ADCK5 | |
0.1 mL | |
Protein Kinase | |
203054 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction