Learn More
Memmert™ Additional Stainless Steel Grid for UF160plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ UF160plusE29767
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
UF160plus Oven | |
Reinforced Grid | |
Stainless Steel | |
60 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.