accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Adhesive Polyethylene Fire Extinguisher Pictograms
Print
Brady™ Adhesive Polyethylene Fire Extinguisher Pictograms
61.50 SEK - 150.95 SEK
Products
2
Documents
Product Certifications
Products
2
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
Product Code
Brand
Dimensions (L x W)
Price
Quantity & Availability
11760564
View Documents
missing translation for 'viewProductCertificates'
Brady™
251127
L x H: 200 x 250 mm
61.50 SEK
Each
Estimated Shipment: 12-12-2024
Log in
to see stock.
Add to basket
11730853
View Documents
missing translation for 'viewProductCertificates'
Brady™
807687
L x H: 315 x 315 mm
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Documents
Product Certifications
Back to Top