missing translation for 'onlineSavingsMsg'
Learn More

AHSA1 Rabbit anti-Human, Mouse, Rat, Clone: 8E5H7, Novus Biologicals™

Rabbit Monoclonal Antibody

1905.00 SEK - 4805.00 SEK

Specifications

Antigen AHSA1
Clone 8E5H7
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18384984
View Documents
Bio-Techne
NBP3-16820-20UL
20 μg
1905.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18321715
View Documents
Bio-Techne
NBP3-16820-100UL
100 μg
4805.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AHSA1 Monoclonal antibody specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

AHSA1
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS, 0.05% BSA, 50% glycerol, pH7.3
10598
IgG
Affinity purified
8E5H7
Monoclonal
Purified
RUO
Human, Mouse, Rat
activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
A synthetic peptide corresponding to a sequence within amino acids 239-338 of human AHSA1 (O95433).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.