missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
1654.00 SEK - 4180.00 SEK
Specifications
Antigen | AIM1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17763654
|
Invitrogen
PA5122258 |
20 μL |
1654.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17753654
|
Invitrogen
PA5122257 |
100 μL |
4180.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIM1 Polyclonal antibody specifically detects AIM1 in Human samples. It is validated for Western Blot.Specifications
AIM1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y4K1 | |
202 | |
Pro1502-Met1719 (Accession Q9Y4K1), with N-terminal His Tag | |
Primary | |
-20°C | |
CRYBG1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
absent in melanoma 1; absent in melanoma 1 protein; AIM1; beta/gamma crystallin domain-containing protein 1; beta-gamma crystallin domain containing 1; CRYBG1; ST4; suppression of tumorigenicity 4 (malignant melanoma) | |
CRYBG1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |