Learn More
Abeba™ Air-Cushion 32600 Shoes
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
Brand:  Abeba™ 32600/44
 View more versions of this product
Product Code. 15639727
  • 1045.00 SEK / 1 pair
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Description
  • CE EN ISO 20347:2012 O2 SRA
  • Grained leather
  • White
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 SRA
Microfiber
44
Polyurethane
White
Leather
Slip-resistant
32600