Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
Brand:  Notrax 410C0324GY
 View more versions of this product
Product Code. 17752058
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Ribbed pattern enhances traction
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Specifications
Gray
1830 cm
Industrial and Commercial Matting
60 ft.
Microcellular Vinyl
60 cm