missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1B1 Mouse anti-Human, Clone: rAKR1B1/7295, Novus Biologicals™
Mouse Monoclonal Antibody
2470.00 SEK - 4910.00 SEK
Specifications
Antigen | AKR1B1 |
---|---|
Clone | rAKR1B1/7295 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18389623
|
Bio-Techne
NBP3-13895-20UG |
20 μg |
2470.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323133
|
Bio-Techne
NBP3-13895-100UG |
100 μg |
4910.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKR1B1 Monoclonal antibody specifically detects AKR1B1 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
AKR1B1 | |
0.2 mg/ml | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
ADR, Aldehyde reductase, Aldo-keto reductase family 1 member B1, aldo-keto reductase family 1, member B1 (aldose reductase), ALDR1aldose reductase, ALR2, ARaldehyde reductase 1, EC 1.1.1, EC 1.1.1.21, Lii5-2 CTCL tumor antigen, low Km aldose reductase, MGC1804 | |
Recombinant human full-length AKR1B1 protein (Uniprot: P15121) | |
Primary | |
Store at 4°C. |
rAKR1B1/7295 | |
Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P15121 | |
231 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title