missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT2 Monoclonal Antibody (ZA006), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 393900
This item is not returnable.
View return policy
Description
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone. Akt is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.Specifications
AKT2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P31751, P47197 | |
Akt2 | |
Synthetic peptide derived from the C-terminal region of the human Akt2 protein, which is 95% and 90% homologous with rat and mouse, respectively | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
ZA006 | |
Unconjugated | |
Akt2 | |
2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 | |
Mouse | |
Protein A | |
RUO | |
208, 25233 | |
-20°C | |
Liquid |