Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2.7mm, I.D.: 1.7mm, Thermo Scientific Chemicals
POA - 647.00 SEK
Specifications
Chemical Name or Material | Al-23 Insulating Tube |
---|---|
Name Note | Both ends open (Thin Wall) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353897
|
Thermo Scientific Alfa Aesar
031492.DM |
100 mm | N/A | ||||||
11363897
|
Thermo Scientific Alfa Aesar
031492.DR |
200 mm |
562.10 SEK
200mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343897
|
Thermo Scientific Alfa Aesar
031492.DW |
300 mm | N/A | ||||||
15401197
|
Thermo Scientific Alfa Aesar
031492.Q@ |
400 mm | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Al-23 Insulating Tube | |
Both ends open (Thin Wall) |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
RUO – Research Use Only