Learn More
Al-24 Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Thermo Scientific Chemicals
2425.00 SEK - 4996.00 SEK
Specifications
Chemical Name or Material Al-24 Tube
Name Note One end closed
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11372868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.DR
200 mm
3171.45 SEK
200mm
View alternative products
Save Up to 
»
11362868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.DW
300 mm
3254.28 SEK
300mm
View alternative products
Save Up to 
»
11332868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11312868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11342868
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030093.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-24 Tube
One end closed
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures