Learn More
Aliquat™ 336, average MW 442, Thermo Scientific™
290.19 SEK - 732.51 SEK
Chemical Identifiers
CAS | 63393-96-4 |
---|---|
Molecular Formula | C25H54ClN |
Molecular Weight (g/mol) | 404.16 |
MDL Number | MFCD00011862 |
InChI Key | XKBGEWXEAPTVCK-UHFFFAOYSA-M |
Synonym | methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride |
PubChem CID | 21218 |
ChEBI | CHEBI:75286 |
IUPAC Name | methyltrioctylazanium chloride |
SMILES | [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Phase transfer catalystChemical Identifiers
63393-96-4 | |
404.16 | |
XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
21218 | |
methyltrioctylazanium chloride |
C25H54ClN | |
MFCD00011862 | |
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
CHEBI:75286 | |
[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC |
Specifications
63393-96-4 | |
0.8800g/mL | |
Authentic | |
1.4655 to 1.4715 | |
05,460; 06,404; 07,380; 13,192 | |
XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
methyltrioctylazanium chloride | |
21218 | |
442 | |
Viscous Liquid |
Amber | |
132°C | |
C25H54ClN | |
MFCD00011862 | |
methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC | |
404.16 | |
CHEBI:75286 | |
88.2 to 90.6%, 112.0 to 115.0 (quaternary value) | |
Aliquat™ 336, Average M.W. 442 |
Safety and Handling
GHS P Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Aliquat]TS 336
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.