missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizarin-3-Methyliminodiacetic Acid, 85%, Honeywell Fluka™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifica
Melting Point | 185°C |
Color | Yellow |
Packaging | Glass Bottle |
Quantity | 5 g |
UN Number | NONH for all modes of transport |
Beilstein | 2190028 |
Formula Weight | 385.32g/mol |
Percent Purity | 85% |
Chemical Name or Material | Alizarin-3-Methyliminodiacetic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'