Learn More
Brady™ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
271.12 SEK - 722.02 SEK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | RoHS Compliant to 2011/65/EU (Pass 2015/863) |
Adhesive Type | Permanent Acrylic |
Coating | None |
Finish | Gloss |
Product Code | Brand | Color | Dimensions (LxW) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | For Use With (Equipment) | Price | Quantity & Availability | |||
17881809
|
Brady™
170949 |
Black on White | 25 ft. x 0.25 in. | BMP41, BMP51, M511 |
271.12 SEK
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17851819
|
Brady™
170950 |
Black on Yellow | 25 ft. x 0.25 in. | BMP41, BMP51, M511 |
271.12 SEK
Each |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17941940
|
Brady™
170845 |
White on Red | 25 ft. x 0.375 in. | BMP41, BMP51, M511 |
361.46 SEK
Each |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17910770
|
Brady™
170843 |
Black on White | 25 ft. x 0.375 in. | BMP41, BMP51, M511 |
361.46 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17940710
|
Brady™
170844 |
Black on Yellow | 25 ft. x 0.375 in. | BMP41, BMP51, M511 |
361.46 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17961960
|
Brady™
170821 |
White on Black | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17910700
|
Brady™
170813 |
White on Blue | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17900700
|
Brady™
170814 |
Black, Clear | 20 ft. x 0.5 in. | BMP41, BMP51, M511 |
391.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17970700
|
Brady™
170822 |
White, Clear | 20 ft. x 0.5 in. | BMP41, BMP51, M511 |
391.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17920670
|
Brady™
170815 |
White on Green | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17970760
|
Brady™
170816 |
Black on Orange | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17930700
|
Brady™
170817 |
White on Red | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17950700
|
Brady™
170818 |
Black on White | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17920700
|
Brady™
170819 |
Red on White | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17940670
|
Brady™
170820 |
Black on Yellow | 25 ft. x 0.5 in. | BMP41, BMP51, M511 |
481.50 SEK
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17941960
|
Brady™
170823 |
White on Black | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17921960
|
Brady™
170824 |
White on Blue | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17901940
|
Brady™
170825 |
Black, Clear | 20 ft. x 0.75 in. | BMP41, BMP51, M511 |
440.21 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17971960
|
Brady™
170826 |
White, Clear | 20 ft. x 0.75 in. | BMP41, BMP51, M511 |
440.21 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17931950
|
Brady™
170827 |
White on Green | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17921950
|
Brady™
170828 |
Black on Orange | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17941950
|
Brady™
170829 |
White on Red | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17981950
|
Brady™
170830 |
Black on White | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17951950
|
Brady™
170831 |
Red on White | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17951960
|
Brady™
170832 |
Black on Yellow | 25 ft. x 0.75 in. | BMP41, BMP51, M511 |
541.24 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17960670
|
Brady™
170841 |
White on Black | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17930670
|
Brady™
170836 |
White on Blue | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17910690
|
Brady™
170838 |
Black, Clear | 20 ft. x 1 in. | BMP41, BMP51, M511 |
489.15 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17980700
|
Brady™
170842 |
White, Clear | 20 ft. x 1 in. | BMP41, BMP51, M511 |
489.15 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17960690
|
Brady™
170839 |
White on Green | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17960700
|
Brady™
170835 |
Black on Orange | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17930690
|
Brady™
170837 |
White on Red | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17970690
|
Brady™
170833 |
Black on White | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17950690
|
Brady™
170840 |
Red on White | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17940690
|
Brady™
170834 |
Black on Yellow | 25 ft. x 1 in. | BMP41, BMP51, M511 |
601.76 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17891809
|
Brady™
170947 |
White on Black | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17811789
|
Brady™
170939 |
White on Blue | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17891709
|
Brady™
170940 |
Black, Clear | 20 ft. x 1.5 in. | BMP51, M511 |
586.80 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17801819
|
Brady™
170948 |
White, Clear | 20 ft. x 1.5 in. | BMP51, M511 |
586.80 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17821769
|
Brady™
170941 |
White on Green | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17871819
|
Brady™
170942 |
Black on Orange | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17811769
|
Brady™
170943 |
White on Red | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17821719
|
Brady™
170944 |
Black on White | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17861819
|
Brady™
170945 |
Red on White | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17861829
|
Brady™
170946 |
Black on Yellow | 25 ft. x 1.5 in. | BMP51, M511 |
722.02 SEK
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Ideal for labeling pipes, equipment, shelves and other industrial surfaces
- Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
- Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
- Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Gloss | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
1 Tape/Cartridge | |
0.004 in. |
RoHS Compliant to 2011/65/EU (Pass 2015/863) | |
None | |
Continuous | |
Top | |
Labels | |
Component and Equipment Labeling, Electrical Labeling, Facility Identification, Floor Marking, Laboratory Labeling, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking | |
-40°F to +180°F | |
0.5 in. |